Learn More
Biomatik Corporation Human C1q And Tumor Necrosis Factor Related Protein 3 (C1QTNF3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02829
Encompass_Preferred
The Human C1q And Tumor Necrosis Factor Related Protein 3 (C1QTNF3) ELISA Kit allows for quantitative determination of C1q And Tumor Necrosis Factor Related Protein 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BXJ4. Cross Activity: No significant cross-reactivity or interference between C1q And Tumor Necrosis Factor Related Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1973