Learn More
Biomatik Corporation Human C1q And Tumor Necrosis Factor Related Protein 9 (C1QTNF9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02830
Encompass_Preferred
The Human C1q And Tumor Necrosis Factor Related Protein 9 (C1QTNF9) ELISA Kit allows for quantitative determination of C1q And Tumor Necrosis Factor Related Protein 9 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P0C862. Cross Activity: No significant cross-reactivity or interference between C1q And Tumor Necrosis Factor Related Protein 9 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1974