Learn More
Biomatik Corporation Rat Cadherin 5 (CDH5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02838
Encompass_Preferred
Rat Cadherin 5 (CDH5) ELISA Kit for detecting CDH5 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1M7E5. Target Synonyms: CD144; 7B4; Cadherin 5 Type 2; VE-Cadherin; Cadherin,Vascular Endothelial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1979