Learn More
Biomatik Corporation Human Cadherin, Epithelial (CDHE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02841
Encompass_Preferred
The Human Cadherin, Epithelial (CDHE) ELISA Kit allows for quantitative determination of Cadherin, Epithelial in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 25.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12830. Cross Activity: No significant cross-reactivity or interference between Cadherin, Epithelial and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1980