Learn More
Biomatik Corporation Human Cadherin, Osteoblast (CDHOB) ELISA Kit, 96 tests
Catalog No. 501501982
Encompass_Preferred
The Human Cadherin, Osteoblast (CDHOB) ELISA Kit allows for quantitative determination of Cadherin, Osteoblast in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55287. Cross Activity: No significant cross-reactivity or interference between Cadherin, Osteoblast and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1982 Supplier Biomatik Corporation Supplier No. EKU02846
The Human Cadherin, Osteoblast (CDHOB) ELISA Kit allows for quantitative determination of Cadherin, Osteoblast in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55287. Cross Activity: No significant cross-reactivity or interference between Cadherin, Osteoblast and analogues was observed. Restriction: For Research Use Only.