Learn More
Biomatik Corporation Dog Calcitonin (CT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02852
Encompass_Preferred
The Dog Calcitonin (CT) ELISA Kit allows for quantitative determination of Calcitonin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 12.35-1000 pg/mL. Sensitivity: 5.28 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P41547. Cross Activity: No significant cross-reactivity or interference between Calcitonin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1985