Learn More
Biomatik Corporation Human Calcitonin Gene Related Peptide (CGRP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02856
Encompass_Preferred
The Human Calcitonin Gene Related Peptide (CGRP) ELISA Kit allows for quantitative determination of Calcitonin Gene Related Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 5.35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P10092. Cross Activity: No significant cross-reactivity or interference between Calcitonin Gene Related Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1989