Learn More
Biomatik Corporation Mouse Calcitonin Gene Related Peptide (CGRP) ELISA Kit, 96 tests
Catalog No. 501501990
Encompass_Preferred
Mouse Calcitonin Gene Related Peptide (CGRP) ELISA Kit for detecting CGRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q99MP3. Target Synonyms: CALCB; CGRP2; CALC2; CGRP-II; Calcitonin Gene Related Peptide 2; Calcitonin 2; Calcitonin Related Polypeptide Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1990 Supplier Biomatik Corporation Supplier No. EKU02857
Mouse Calcitonin Gene Related Peptide (CGRP) ELISA Kit for detecting CGRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q99MP3. Target Synonyms: CALCB; CGRP2; CALC2; CGRP-II; Calcitonin Gene Related Peptide 2; Calcitonin 2; Calcitonin Related Polypeptide Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.