Learn More
Biomatik Corporation Human Calpain 3 (CAPN3) ELISA Kit, 96 tests
Catalog No. 501501994
Encompass_Preferred
The Human Calpain 3 (CAPN3) ELISA Kit allows for quantitative determination of Calpain 3 in Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P20807. Cross Activity: No significant cross-reactivity or interference between Calpain 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1994 Supplier Biomatik Corporation Supplier No. EKU02886
The Human Calpain 3 (CAPN3) ELISA Kit allows for quantitative determination of Calpain 3 in Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P20807. Cross Activity: No significant cross-reactivity or interference between Calpain 3 and analogues was observed. Restriction: For Research Use Only.