Learn More
Biomatik Corporation Human Calprotectin (CALPRO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02892
Encompass_Preferred
The Human Calprotectin (CALPRO) ELISA Kit allows for quantitative determination of Calprotectin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05109. Cross Activity: No significant cross-reactivity or interference between Calprotectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1995