Learn More
Biomatik Corporation Human Carbonic Anhydrase II (CA2) ELISA Kit, 96 tests
Catalog No. 501502004
Encompass_Preferred
The Human Carbonic Anhydrase II (CA2) ELISA Kit allows for quantitative determination of Carbonic Anhydrase II in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00918. Cross Activity: No significant cross-reactivity or interference between Carbonic Anhydrase II and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2004 Supplier Biomatik Corporation Supplier No. EKU02918
The Human Carbonic Anhydrase II (CA2) ELISA Kit allows for quantitative determination of Carbonic Anhydrase II in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00918. Cross Activity: No significant cross-reactivity or interference between Carbonic Anhydrase II and analogues was observed. Restriction: For Research Use Only.