Learn More
Biomatik Corporation Human Carbonic Anhydrase VI (CA6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02926
Encompass_Preferred
The Human Carbonic Anhydrase VI (CA6) ELISA Kit allows for quantitative determination of Carbonic Anhydrase VI in Serum, Plasma, Saliva and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.131 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23280. Cross Activity: No significant cross-reactivity or interference between Carbonic Anhydrase VI and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2006