Learn More
Biomatik Corporation Human Carbonic Anhydrase XII (CA12) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02927
Encompass_Preferred
The Human Carbonic Anhydrase XII (CA12) ELISA Kit allows for quantitative determination of Carbonic Anhydrase XII in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43570. Cross Activity: No significant cross-reactivity or interference between Carbonic Anhydrase XII and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2007