Learn More
Biomatik Corporation Rat Carboxypeptidase B1, Tissue (CPB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02937
Encompass_Preferred
Rat Carboxypeptidase B1, Tissue (CPB1) ELISA Kit for detecting CPB1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19223. Target Synonyms: CPB; PCPB; PASP; Pancreas-specific protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2009