Learn More
Biomatik Corporation Human Carboxypeptidase E (CPE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02938
Encompass_Preferred
The Human Carboxypeptidase E (CPE) ELISA Kit allows for quantitative determination of Carboxypeptidase E in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.236 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16870. Cross Activity: No significant cross-reactivity or interference between Carboxypeptidase E and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2010