Learn More
Biomatik Corporation Human Carcinoembryonic Antigen (CEA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02944
Encompass_Preferred
The Human Carcinoembryonic Antigen (CEA) ELISA Kit allows for quantitative determination of Carcinoembryonic Antigen in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 19.6-1250 pg/mL. Sensitivity: 7.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06731. Cross Activity: No significant cross-reactivity or interference between Carcinoembryonic Antigen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2012