Learn More
Biomatik Corporation Mouse Carcinoembryonic Antigen (CEA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02945
Encompass_Preferred
Mouse Carcinoembryonic Antigen (CEA) ELISA Kit for detecting CEA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3UKK2. Gene ID: 73250. Target Synonyms: CD66E; CD66; CEACAM5; Carcinoembryonic Antigen-related Cell Adhesion Molecule 5; Carcinoembryonic Antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2013