Learn More
Biomatik Corporation Human Cardiotrophin 1 (CT1) ELISA Kit, 96 tests
Catalog No. 501502015
Encompass_Preferred
The Human Cardiotrophin 1 (CT1) ELISA Kit allows for quantitative determination of Cardiotrophin 1 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q16619. Cross Activity: No significant cross-reactivity or interference between Cardiotrophin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2015 Supplier Biomatik Corporation Supplier No. EKU02952
The Human Cardiotrophin 1 (CT1) ELISA Kit allows for quantitative determination of Cardiotrophin 1 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q16619. Cross Activity: No significant cross-reactivity or interference between Cardiotrophin 1 and analogues was observed. Restriction: For Research Use Only.