Learn More
Biomatik Corporation Mouse Carnitine Palmitoyltransferase 1A, Liver (CPT1A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02962
Encompass_Preferred
Mouse Carnitine Palmitoyltransferase 1A, Liver (CPT1A) ELISA Kit for detecting CPT1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97742. Gene ID: 12894. Target Synonyms: CPT1-L; Carnitine O-palmitoyltransferase I, liver isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2018