Learn More
Biomatik Corporation Rat Carnitine Palmitoyltransferase 1A, Liver (CPT1A) ELISA Kit, 96 tests
Catalog No. 501502019
Encompass_Preferred
Rat Carnitine Palmitoyltransferase 1A, Liver (CPT1A) ELISA Kit for detecting CPT1A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32198. Gene ID: 25757. Target Synonyms: CPT1-L; Carnitine O-palmitoyltransferase I, liver isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2019 Supplier Biomatik Corporation Supplier No. EKU02963
Rat Carnitine Palmitoyltransferase 1A, Liver (CPT1A) ELISA Kit for detecting CPT1A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32198. Gene ID: 25757. Target Synonyms: CPT1-L; Carnitine O-palmitoyltransferase I, liver isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.