Learn More
Biomatik Corporation Human Cartilage Oligomeric Matrix Protein (COMP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02973
Encompass_Preferred
The Human Cartilage Oligomeric Matrix Protein (COMP) ELISA Kit allows for quantitative determination of Cartilage Oligomeric Matrix Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49747. Cross Activity: No significant cross-reactivity or interference between Cartilage Oligomeric Matrix Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2022