Learn More
Biomatik Corporation Mouse Cartilage Oligomeric Matrix Protein (COMP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02974
Encompass_Preferred
Mouse Cartilage Oligomeric Matrix Protein (COMP) ELISA Kit for detecting COMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R0G6. Gene ID: 12845. Target Synonyms: MED; EDM1; EPD1; PSACH; THBS5; TSP-5; Pseudoachondroplasia,Epiphyseal Dysplasia 1,Multiple; Thrombospondin 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2023