Learn More
Biomatik Corporation Cattle Casein Alpha (CSN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02976
Encompass_Preferred
The Cattle Casein Alpha (CSN1) ELISA Kit allows for quantitative determination of Casein Alpha in milk. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.068 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02662. Cross Activity: No significant cross-reactivity or interference between Casein Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2025