Learn More
Biomatik Corporation Human Casein Alpha (CSN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02977
Encompass_Preferred
The Human Casein Alpha (CSN1) ELISA Kit allows for quantitative determination of Casein Alpha in Tissue homogenates, Cell lysates, Cell culture supernates, Breast milk and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P47710. Cross Activity: No significant cross-reactivity or interference between Casein Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2026