Learn More
Biomatik Corporation Human Casein Kappa (CSN3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02981
Encompass_Preferred
The Human Casein Kappa (CSN3) ELISA Kit allows for quantitative determination of Casein Kappa in Breast milk and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07498. Cross Activity: No significant cross-reactivity or interference between Casein Kappa and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2030