Learn More
Biomatik Corporation Mouse Caspase 1 (CASP1) ELISA Kit, 96 tests
Catalog No. 501502033
Encompass_Preferred
Mouse Caspase 1 (CASP1) ELISA Kit for detecting CASP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29452. Gene ID: 12362. Target Synonyms: ICE; IL1BC; IL1-BC; P45; Interleukin 1 Beta Converting Enzyme; Apoptosis-Related Cysteine Peptidase; Interleukin-1 beta convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2033 Supplier Biomatik Corporation Supplier No. EKU02984
Mouse Caspase 1 (CASP1) ELISA Kit for detecting CASP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29452. Gene ID: 12362. Target Synonyms: ICE; IL1BC; IL1-BC; P45; Interleukin 1 Beta Converting Enzyme; Apoptosis-Related Cysteine Peptidase; Interleukin-1 beta convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.