Learn More
Biomatik Corporation Rat Caspase 1 (CASP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02985
Encompass_Preferred
Rat Caspase 1 (CASP1) ELISA Kit for detecting CASP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43527. Target Synonyms: ICE; IL1BC; IL1-BC; P45; Interleukin 1 Beta Converting Enzyme; Apoptosis-Related Cysteine Peptidase; Interleukin-1 beta convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2034