Learn More
Biomatik Corporation Human Caspase 8 (CASP8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02998
Encompass_Preferred
The Human Caspase 8 (CASP8) ELISA Kit allows for quantitative determination of Caspase 8 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.244 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14790. Cross Activity: No significant cross-reactivity or interference between Caspase 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2040