Learn More
Biomatik Corporation Human Cat Eye Syndrome Chromosome Region, Candidate 1 (CECR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03006
Encompass_Preferred
The Human Cat Eye Syndrome Chromosome Region, Candidate 1 (CECR1) ELISA Kit allows for quantitative determination of Cat Eye Syndrome Chromosome Region, Candidate 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NZK5. Cross Activity: No significant cross-reactivity or interference between Cat Eye Syndrome Chromosome Region, Candidate 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2042