Learn More
Biomatik Corporation Rat Catenin Beta 1 (CTNNb1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03016
Encompass_Preferred
Rat Beta Catenin (β-catenin) ELISA Kit for detecting β-catenin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WU82. Gene ID: 84353. Target Synonyms: CTNNb1; bCat; b-Cat; CTNN-B1; Catenin Beta 1; Cadherin-Associated Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2047