Learn More
Biomatik Corporation Human Cathepsin L (CTSL) ELISA Kit, 96 tests
Catalog No. 501502056
Encompass_Preferred
The Human Cathepsin L (CTSL) ELISA Kit allows for quantitative determination of Cathepsin L in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 25.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07711. Cross Activity: No significant cross-reactivity or interference between Cathepsin L and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2056 Supplier Biomatik Corporation Supplier No. EKU03037
The Human Cathepsin L (CTSL) ELISA Kit allows for quantitative determination of Cathepsin L in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 25.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07711. Cross Activity: No significant cross-reactivity or interference between Cathepsin L and analogues was observed. Restriction: For Research Use Only.