Learn More
Biomatik Corporation Rat Cathepsin S (CTSS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03042
Encompass_Preferred
Rat Cathepsin S (CTSS) ELISA Kit for detecting CTSS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 23.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02765. Gene ID: 50654. Target Synonyms: CTS-S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2060