Learn More
Biomatik Corporation Human CD5 Antigen Like Protein (CD5L) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03060
Encompass_Preferred
The Human CD5 Antigen Like Protein (CD5L) ELISA Kit allows for quantitative determination of CD5 Antigen Like Protein in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43866. Cross Activity: No significant cross-reactivity or interference between CD5 Antigen Like Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2065