Learn More
Biomatik Corporation Human Cell Adhesion Molecule With Homology To L1CAM (CHL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03063
Encompass_Preferred
The Human Cell Adhesion Molecule With Homology To L1CAM (CHL1) ELISA Kit allows for quantitative determination of Cell Adhesion Molecule With Homology To L1CAM in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.060 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00533. Cross Activity: No significant cross-reactivity or interference between Cell Adhesion Molecule With Homology To L1CAM and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2066