Learn More
Biomatik Corporation Human Cellular Retinoic Acid Binding Protein 2 (CRABP2) ELISA Kit, 96 tests
Catalog No. 501502067
Encompass_Preferred
The Human Cellular Retinoic Acid Binding Protein 2 (CRABP2) ELISA Kit allows for quantitative determination of Cellular Retinoic Acid Binding Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29373. Cross Activity: No significant cross-reactivity or interference between Cellular Retinoic Acid Binding Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2067 Supplier Biomatik Corporation Supplier No. EKU03069
The Human Cellular Retinoic Acid Binding Protein 2 (CRABP2) ELISA Kit allows for quantitative determination of Cellular Retinoic Acid Binding Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29373. Cross Activity: No significant cross-reactivity or interference between Cellular Retinoic Acid Binding Protein 2 and analogues was observed. Restriction: For Research Use Only.