Learn More
Biomatik Corporation Human Ceramide Kinase (CERK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03073
Encompass_Preferred
The Human Ceramide Kinase (CERK) ELISA Kit allows for quantitative determination of Ceramide Kinase in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.14 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8TCT0. Cross Activity: No significant cross-reactivity or interference between Ceramide Kinase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2068