Learn More
Biomatik Corporation Rat Cerebral Dopamine Neurotrophic Factor (CDNF) ELISA Kit, 96 tests
Catalog No. 501502071
Encompass_Preferred
Rat Cerebral Dopamine Neurotrophic Factor (CDNF) ELISA Kit for detecting CDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0C5I0. Gene ID: 361276. Target Synonyms: ARMETL1; ARMET-like protein 1; Conserved Dopamine Neurotrophic Factor; Arginine-Rich,Mutated In Early Stage Tumors-Like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2071 Supplier Biomatik Corporation Supplier No. EKU03080
Rat Cerebral Dopamine Neurotrophic Factor (CDNF) ELISA Kit for detecting CDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0C5I0. Gene ID: 361276. Target Synonyms: ARMETL1; ARMET-like protein 1; Conserved Dopamine Neurotrophic Factor; Arginine-Rich,Mutated In Early Stage Tumors-Like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.