Learn More
Biomatik Corporation Mouse Ceruloplasmin (CP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03082
Encompass_Preferred
Mouse Ceruloplasmin (CP) ELISA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61147. Gene ID: 12870. Target Synonyms: CP2; CER; Caeruloplasmin; Ferroxidase; Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2073