Learn More
Biomatik Corporation Human Chemerin (CHEM) ELISA Kit, 96 tests
Catalog No. 501502075
Encompass_Preferred
The Human Chemerin (CHEM) ELISA Kit allows for quantitative determination of Chemerin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.069 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q99969. Cross Activity: No significant cross-reactivity or interference between Chemerin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2075 Supplier Biomatik Corporation Supplier No. EKU03089
The Human Chemerin (CHEM) ELISA Kit allows for quantitative determination of Chemerin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.069 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q99969. Cross Activity: No significant cross-reactivity or interference between Chemerin and analogues was observed. Restriction: For Research Use Only.