Learn More
Biomatik Corporation Human Chemokine C-C-Motif Ligand 14 (CCL14) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03094
The Human Chemokine C-C-Motif Ligand 14 (CCL14) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Ligand 14 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q16627. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Ligand 14 and analogues was observed. Restriction: For Research Use Only.