Learn More
Biomatik Corporation Human Chemokine C-C-Motif Receptor 5 (CCR5) ELISA Kit, 96 tests
Catalog No. 501502079
Encompass_Preferred
The Human Chemokine C-C-Motif Receptor 5 (CCR5) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Receptor 5 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51681. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Receptor 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2079 Supplier Biomatik Corporation Supplier No. EKU03103
The Human Chemokine C-C-Motif Receptor 5 (CCR5) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Receptor 5 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51681. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Receptor 5 and analogues was observed. Restriction: For Research Use Only.