Learn More
Biomatik Corporation Human Chemokine C-C-Motif Receptor 6 (CCR6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03104
Encompass_Preferred
The Human Chemokine C-C-Motif Receptor 6 (CCR6) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Receptor 6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51684. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Receptor 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2080