Learn More
Biomatik Corporation Human Chemokine C-C-Motif Receptor 7 (CCR7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03106
Encompass_Preferred
The Human Chemokine C-C-Motif Receptor 7 (CCR7) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Receptor 7 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P32248. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Receptor 7 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2081
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.