Learn More
Biomatik Corporation Mouse Chemokine C-X3-C-Motif Ligand 1 (CX3CL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03110
Encompass_Preferred
Mouse Chemokine C-X3-C-Motif Ligand 1 (CX3CL1) ELISA Kit for detecting CX3CL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35188. Gene ID: 20312. Target Synonyms: NTN; ABCD3; C3Xkine; CXC3; CXC3C; NTT; SCYD1; ABCD3; FKN; Neurotactin; Fractalkine; Small Inducible Cytokine Subfamily D (Cys-X3-Cys) Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2083