Learn More
Biomatik Corporation Human Chloride Channel Accessory 1 (CLCA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03128
Encompass_Preferred
The Human Chloride Channel Accessory 1 (CLCA1) ELISA Kit allows for quantitative determination of Chloride Channel Accessory 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: A8K7I4. Cross Activity: No significant cross-reactivity or interference between Chloride Channel Accessory 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2093