Learn More
Biomatik Corporation Rat Cholecystokinin (CCK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03135
Encompass_Preferred
Rat Cholecystokinin (CCK) ELISA Kit for detecting CCK in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.88-800pg/ml. Sensitivity: 4.22pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01355. Gene ID: 25298. Target Synonyms: CCK-PZ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2098