Learn More
Biomatik Corporation Human Cholesteryl Ester Transfer Protein (CETP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03152
Encompass_Preferred
The Human Cholesteryl Ester Transfer Protein (CETP) ELISA Kit allows for quantitative determination of Cholesteryl Ester Transfer Protein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11597. Cross Activity: No significant cross-reactivity or interference between Cholesteryl Ester Transfer Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2100