Learn More
Biomatik Corporation Mouse Choline Acetyltransferase (ChAT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03155
Encompass_Preferred
Mouse Choline Acetyltransferase (ChAT) ELISA Kit for detecting ChAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03059. Target Synonyms: CMS1A; CMS1A2; CHOACTase; Choline acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2102