Learn More
Biomatik Corporation Rat Cholinergic Receptor, Nicotinic, Alpha 1 (CHRNa1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03162
Encompass_Preferred
Rat Cholinergic Receptor, Nicotinic, Alpha 1 (CHRNa1) ELISA Kit for detecting CHRNa1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25108. Gene ID: 79557. Target Synonyms: CHR-NA1; N-AChRA1; NAChRA1; ACHRA; ACHRD; CMS2A; FCCMS; SCCMS; Acetylcholine receptor subunit alpha; N-AChR-A1; Neuronal Acetylcholine Receptor Alpha 1,Muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2104