Learn More
Biomatik Corporation Human Cholinergic Receptor, Nicotinic, Alpha 7 (CHRNa7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03165
Encompass_Preferred
The Human Cholinergic Receptor, Nicotinic, Alpha 7 (CHRNa7) ELISA Kit allows for quantitative determination of Cholinergic Receptor, Nicotinic, Alpha 7 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P36544. Cross Activity: No significant cross-reactivity or interference between Cholinergic Receptor, Nicotinic, Alpha 7 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2105